missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoyl chloride, 98%

CAS: 122-04-3 | C7H4ClNO3 | 185.56 g/mol

Supplier:  Thermo Scientific Chemicals A125430B

 View more versions of this product

Catalog No. AAA125430B


Only null left
Explore available promotions

Description

Description

4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
Specifications

Specifications

4-Nitrobenzoyl chloride
71°C to 74°C
155°C (15 mmHg)
Characteristic
MFCD00007345
UN3261
Moisture sensitive
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
SKDHHIUENRGTHK-UHFFFAOYSA-N
4-nitrobenzoyl chloride
8502
98%
122-04-3
1.53
102°C (215°F)
C7H4ClNO3
1000 g
473192
14,6590
Soluble in terahydrofuran,dichloromethane,chloroform and pyridine.
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
185.56
185.57

Chemical Identifiers

122-04-3
185.56
SKDHHIUENRGTHK-UHFFFAOYSA-N
8502
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
C7H4ClNO3
MFCD00007345
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
4-nitrobenzoyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 204-517-4

RTECSNumber : DM6651000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only