missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoyl chloride, 98%

Catalog No. p-7020550
Change view
Click to view available options
Quantity:
10 g
250 g
1000 g
Catalog No. Quantity
AAA1254309 10 g
AAA1254330 250 g
AAA125430B 1000 g
3 options

Catalog No. AAA1254309

Supplier: Thermo Scientific Chemicals A1254309

Only null left

CAS: 122-04-3 | C7H4ClNO3 | 185.56 g/mol

4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.

Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.

Chemical Identifiers

CAS 122-04-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007345
InChI Key SKDHHIUENRGTHK-UHFFFAOYSA-N
Synonym p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride
PubChem CID 8502
IUPAC Name 4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 71°C to 74°C
Density 1.53
Boiling Point 155°C (15 mmHg)
Flash Point 102°C (215°F)
Odor Characteristic
Quantity 10 g
UN Number UN3261
Beilstein 473192
Sensitivity Moisture sensitive
Merck Index 14,6590
Solubility Information Soluble in terahydrofuran,dichloromethane,chloroform and pyridine.
Formula Weight 185.57
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzoyl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-517-4
RTECSNumber DM6651000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only