missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzotrifluoride, 98%

Catalog No. AAL1576606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1576606 5 g
AAL1576603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1576606 Supplier Thermo Scientific Chemicals Supplier No. L1576606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 402-54-0 | C7H4F3NO2 | 191.109 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-54-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD00007358
InChI Key XKYLCLMYQDFGKO-UHFFFAOYSA-N
Synonym 4-nitrobenzotrifluoride, 1-nitro-4-trifluoromethyl benzene, p-nitrobenzotrifluoride, benzene, 1-nitro-4-trifluoromethyl, 4-nitro-alpha,alpha,alpha-trifluorotoluene, 4-trifluoromethyl nitrobenzene, 4-nitro-a,a,a-trifluorotoluene, p-nitro-trifluoromethylphenol, p-trifluoromethylnitrobenzene, 4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene
PubChem CID 9821
IUPAC Name 1-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 38°C to 42°C
Boiling Point 81°C to 83°C (10 mmHg)
Flash Point 88°C (190°F)
Odor Characteristic
Quantity 5 g
UN Number UN3431
Formula Weight 191.11
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzotrifluoride
Hazard Category H302-H315-H319-H330-H335
Hazard Statement GHS H Statement
H301-H311-H330-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZOTRIFLUORIDES, SOLID
EINECSNumber 206-948-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only