missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzophenone 99.0+%, TCI America™

Catalog No. N036525G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N036525G 25 g
N036510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N036525G Supplier TCI America Supplier No. N036525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1144-74-7
Molecular Formula C13H9NO3
Molecular Weight (g/mol) 227.22
MDL Number MFCD00007354
InChI Key ZYMCBJWUWHHVRX-UHFFFAOYSA-N
Synonym 4-nitrobenzophenone, p-nitrobenzophenone, 4-nitrophenyl phenyl methanone, benzophenone, 4-nitro, methanone, 4-nitrophenyl phenyl, 4-nitrophenyl phenyl ketone, 4-nitrophenyl-phenylmethanone, 4-nitrophenyl-phenyl-methanone, 4-nitro-benzophenone
PubChem CID 70839
IUPAC Name (4-nitrophenyl)(phenyl)methanone
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 138°C
Quantity 25 g
Formula Weight 227.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzophenone