missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzophenone, 99%

Catalog No. aab2040514
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAB2040514 25 g
AAB2040506 5 g
2 options

Catalog No. AAB2040514

Supplier: Thermo Scientific Chemicals B2040514

Only null left

CAS: 1144-74-7 | C13H9NO3 | 227.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1144-74-7
Molecular Formula C13H9NO3
Molecular Weight (g/mol) 227.22
MDL Number MFCD00007354
InChI Key ZYMCBJWUWHHVRX-UHFFFAOYSA-N
Synonym 4-nitrobenzophenone, p-nitrobenzophenone, 4-nitrophenyl phenyl methanone, benzophenone, 4-nitro, methanone, 4-nitrophenyl phenyl, 4-nitrophenyl phenyl ketone, 4-nitrophenyl-phenylmethanone, 4-nitrophenyl-phenyl-methanone, 4-nitro-benzophenone
PubChem CID 70839
IUPAC Name (4-nitrophenyl)-phenylmethanone
SMILES [O-][N+](=O)C1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 136°C to 138°C
Quantity 25 g
Beilstein 1912753
Formula Weight 227.22
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzophenone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-542-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only