missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoic Acid 99.0+%, TCI America™

Catalog No. N015625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015625G 25 g
N0156500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N015625G Supplier TCI America Supplier No. N015625G

Chemical Identifiers

CAS 62-23-7
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007352
InChI Key OTLNPYWUJOZPPA-UHFFFAOYSA-N
Synonym 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
PubChem CID 6108
ChEBI CHEBI:262350
SMILES C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 240°C
Color Yellow
Quantity 25 g
Formula Weight 167.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzoic Acid
EINECSNumber (3)-1505
RTECSNumber DH5075000
TSCA Yes