missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzoic acid, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | 239.0°C to 242.0°C |
| Color | Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (GC) |
| Packaging | Plastic bottle |
| Linear Formula | O2NC6H4CO2H |
| Quantity | 1 kg |
| Beilstein | 09,389 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. |
| EINECSNumber | 200-526-2 |
| RTECSNumber | DH5075000 |
| TSCA | TSCA |
RUO – Research Use Only