Learn More
4-Nitrobenzoic acid, 99%
CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol
Supplier: Thermo Scientific Chemicals A147380B
Description
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.
Solubility
Very slightly soluble in cold water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications
| 4-Nitrobenzoic acid | |
| 239°C to 241°C | |
| C7H5NO4 | |
| 1000 g | |
| 14,6589 | |
| Very slightly soluble in cold water. | |
| C1=CC(=CC=C1C(=O)O)[N+](=O)[O-] | |
| 6108 | |
| 167.12 |
| 62-23-7 | |
| 1.61 | |
| MFCD00007352 | |
| 973593 | |
| 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova | |
| OTLNPYWUJOZPPA-UHFFFAOYSA-N | |
| 167.12 | |
| CHEBI:262350 | |
| 99% |
Chemical Identifiers
| 62-23-7 | |
| 167.12 | |
| OTLNPYWUJOZPPA-UHFFFAOYSA-N | |
| 6108 | |
| C1=CC(=CC=C1C(=O)O)[N+](=O)[O-] |
| C7H5NO4 | |
| MFCD00007352 | |
| 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova | |
| CHEBI:262350 |
Safety and Handling
GHS H Statement
H301-H351-H315-H319-H335
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
EINECSNumber : 200-526-2
RTECSNumber : DH5075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only