missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonamide 98.0+%, TCI America™

Catalog No. p-7133943
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06975G 5 g
N069725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06975G Supplier TCI America Supplier No. N06975G
Only null left

Chemical Identifiers

CAS 6325-93-5
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00007937
InChI Key QWKKYJLAUWFPDB-UHFFFAOYSA-N
Synonym benzenesulfonamide, 4-nitro, p-nitrobenzenesulfonamide, 4-nitrobenzolesulfamide, p-nitrophenylsulfonamide, 4-nitrobenzenesulphonamide, benzenesulfonamide, p-nitro, 4-nitro-benzenesulfonamide, unii-tuf52o8y8b, tuf52o8y8b, 4-nitrobenzene-1-sulfonamide
PubChem CID 22784
IUPAC Name 4-nitrobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 182°C
Quantity 5 g
Formula Weight 202.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzenesulfonamide