missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonamide, 97%

Catalog No. AC153020050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153020050 5 g Glass bottle
AC153020250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153020050 Supplier Thermo Scientific Chemicals Supplier No. 153020050

CAS: 6325-93-5 | C6H6N2O4S | 202.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6325-93-5
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00007937
InChI Key QWKKYJLAUWFPDB-UHFFFAOYSA-N
Synonym benzenesulfonamide, 4-nitro, p-nitrobenzenesulfonamide, 4-nitrobenzolesulfamide, p-nitrophenylsulfonamide, 4-nitrobenzenesulphonamide, benzenesulfonamide, p-nitro, 4-nitro-benzenesulfonamide, unii-tuf52o8y8b, tuf52o8y8b, 4-nitrobenzene-1-sulfonamide
PubChem CID 22784
IUPAC Name 4-nitrobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 177.0°C to 182.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4SO2NH2
Quantity 5 g
Beilstein 11, 72
Merck Index 15, 6675
Formula Weight 202.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzenesulfonamide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-691-6
RTECSNumber DB2975000
TSCA TSCA

RUO – Research Use Only