missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzenesulfonamide, 97%

Catalog No. AAA1810106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1810106 5 g
AAA1810114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1810106 Supplier Thermo Scientific Chemicals Supplier No. A1810106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6325-93-5 | C6H6N2O4S | 202.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6325-93-5
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00007937
InChI Key QWKKYJLAUWFPDB-UHFFFAOYSA-N
Synonym benzenesulfonamide, 4-nitro, p-nitrobenzenesulfonamide, 4-nitrobenzolesulfamide, p-nitrophenylsulfonamide, 4-nitrobenzenesulphonamide, benzenesulfonamide, p-nitro, 4-nitro-benzenesulfonamide, unii-tuf52o8y8b, tuf52o8y8b, 4-nitrobenzene-1-sulfonamide
PubChem CID 22784
IUPAC Name 4-nitrobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 178°C to 181°C
Quantity 5 g
Formula Weight 202.19
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzenesulfonamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-691-6
RTECSNumber DB2975000
TSCA Yes

RUO – Research Use Only