missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroanisole, 97%

Catalog No. p-7024459
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1915230 250 g
AAA191520B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1915230 Supplier Thermo Scientific Chemicals Supplier No. A1915230
Only null left

CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-17-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007327
InChI Key BNUHAJGCKIQFGE-UHFFFAOYSA-N
Synonym 4-nitroanisole, p-nitroanisole, 4-methoxynitrobenzene, p-methoxynitrobenzene, benzene, 1-methoxy-4-nitro, nitroanisole, p-nitroanisol, anisole, p-nitro, 4-nitrophenyl methyl ether, unii-g989z7wolh
PubChem CID 7485
ChEBI CHEBI:1911
IUPAC Name 1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 50°C to 54°C
Density 1.233
Boiling Point 260°C
Flash Point 130°C (266°F)
Quantity 250 g
UN Number UN3458
Beilstein 1865361
Merck Index 14,6585
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 4-Nitroanisole
Show More Show Less
Hazard Category H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROANISOLES, SOLID
EINECSNumber 202-825-3
RTECSNumber BZ8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only