missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline, 98%

Catalog No. aaa103690b
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA103690B 1000 g
AAA1036930 250 g
AAA103690I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA103690B Supplier Thermo Scientific Chemicals Supplier No. A103690B
Only null left

CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol

p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

Solubility
Soluble in hot water and alcohol

Notes
Store at 4°C. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 147°C to 149°C
Density 1.437
Boiling Point 260°C (100 mmHg)
Flash Point 165°C (329°F)
Odor Amine-like
Quantity 1000 g
UN Number UN1661
Beilstein 508690
Merck Index 14,6584
Solubility Information Soluble in hot water and alcohol
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 4-Nitroaniline
Show More Show Less
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROANILINES
EINECSNumber 202-810-1
RTECSNumber BY7000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only