missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetanilide, 99%

Catalog No. AC415750250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415750250 25 g Glass bottle
AC415751000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415750250 Supplier Thermo Scientific Chemicals Supplier No. 415750250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 104-04-1 | C8H8N2O3 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-04-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00007303
InChI Key NQRLPDFELNCFHW-UHFFFAOYSA-N
Synonym 4'-nitroacetanilide, n-4-nitrophenyl acetamide, 4-nitroacetanilide, p-nitroacetanilide, acetamide, n-4-nitrophenyl, n-acetyl-4-nitroaniline, p-acetamidonitrobenzene, acetanilide, p-nitro, p-nitrophenylacetanilide, n-acetyl-p-nitroaniline
PubChem CID 7691
IUPAC Name N-(4-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 213°C to 216°C
Color Brown-Green or Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C(O)NHC6H4NO2
Quantity 25 g
Merck Index 15, 6667
Solubility Information Solubility in water: 2.2g/L (25°C)
Formula Weight 180.16
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4′-Nitroacetanilide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-169-

RUO – Research Use Only