missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetanilide, 98%

Catalog No. AAA1353906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1353906 5 g
AAA1353914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1353906 Supplier Thermo Scientific Chemicals Supplier No. A1353906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 104-04-1 | C8H8N2O3 | 180.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-04-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00007303
InChI Key NQRLPDFELNCFHW-UHFFFAOYSA-N
Synonym 4'-nitroacetanilide, n-4-nitrophenyl acetamide, 4-nitroacetanilide, p-nitroacetanilide, acetamide, n-4-nitrophenyl, n-acetyl-4-nitroaniline, p-acetamidonitrobenzene, acetanilide, p-nitro, p-nitrophenylacetanilide, n-acetyl-p-nitroaniline
PubChem CID 7691
IUPAC Name N-(4-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 215°C to 217°C
Quantity 5 g
Beilstein 2211962
Merck Index 14,6581
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 4'-Nitroacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-169-0
RTECSNumber AE5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only