missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-m-cresol 98.0+%, TCI America™

Catalog No. N035125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035125G 25 g
N0351500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N035125G Supplier TCI America Supplier No. N035125G

Chemical Identifiers

CAS 2581-34-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007333
InChI Key PIIZYNQECPTVEO-UHFFFAOYSA-N
Synonym 4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
PubChem CID 17412
ChEBI CHEBI:38683
IUPAC Name 3-methyl-4-nitrophenol
SMILES CC1=CC(O)=CC=C1[N+]([O-])=O

Specifications

Melting Point 128°C
Color Yellow
Quantity 25 g
UN Number 2446
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-m-cresol
EINECSNumber (3)-0790
RTECSNumber GP2625000
TSCA Yes