missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-L-phenylalanine Hydrate 98.0+%, TCI America™

Catalog No. N06821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06821G 1 g
N06825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06821G Supplier TCI America Supplier No. N06821G

Chemical Identifiers

CAS 949-99-5
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.19
MDL Number MFCD00150543
InChI Key GTVVZTAFGPQSPC-QMMMGPOBSA-N
Synonym 4-nitro-l-phenylalanine, h-phe 4-no2-oh, s-2-amino-3-4-nitrophenyl propanoic acid, 4-nitro-3-phenyl-l-alanine, l-4-nitrophenylalanine, p-nitro-l-phenylalanine, 2s-2-amino-3-4-nitrophenyl propanoic acid, p-nitrophenylalanine, l-phenylalanine, 4-nitro, l-4-no2-phe-oh
PubChem CID 13706
IUPAC Name (2S)-2-amino-3-(4-nitrophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Quantity 1 g
Formula Weight 210.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-L-phenylalanine Hydrate
RTECSNumber AY7400000
TSCA No