CAS RN 949-99-5
IUPAC Name:
(2S)-2-amino-3-(4-nitrophenyl)propanoic acid
Synonyms:
phenylalanine, 4-nitro-
Molecular Weight (g/mol):
210.19
Molecular Formula:
C9H10N2O4
InChi Key:
GTVVZTAFGPQSPC-QMMMGPOBSA-N
SMILES:
N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
1
–
1
of
1
results
1
4-Nitro-L-phenylalanine Hydrate 98.0+%, TCI America™
CAS: 949-99-5 Molecular Formula: C9H10N2O4 Molecular Weight (g/mol): 210.19 MDL Number: MFCD00150543 InChI Key: GTVVZTAFGPQSPC-QMMMGPOBSA-N Synonym: 4-nitro-l-phenylalanine,h-phe 4-no2-oh,s-2-amino-3-4-nitrophenyl propanoic acid,4-nitro-3-phenyl-l-alanine,l-4-nitrophenylalanine,p-nitro-l-phenylalanine,2s-2-amino-3-4-nitrophenyl propanoic acid,p-nitrophenylalanine,l-phenylalanine, 4-nitro,l-4-no2-phe-oh PubChem CID: 13706 IUPAC Name: (2S)-2-amino-3-(4-nitrophenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O