missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-D-phenylalanine Hydrate 97.0+%, TCI America™

Catalog No. N08491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08491G 1 g
N08495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08491G Supplier TCI America Supplier No. N08491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56613-61-7
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.19
MDL Number MFCD00150962
InChI Key GTVVZTAFGPQSPC-UHFFFAOYNA-N
Synonym 4-nitro-d-phenylalanine, h-d-phe 4-no2-oh, p-nitro-d-phenylalanine, 2r-2-amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, 4-nitro, r-2-amino-3-4-nitrophenyl propanoic acid, d-4-nitrophe, ambotzhaa1044, pubchem13021, d-4-nitro phenylalanine
PubChem CID 679497
IUPAC Name 2-amino-3-(4-nitrophenyl)propanoic acid
SMILES NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 210.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-D-phenylalanine Hydrate