accessibility menu, dialog, popup

UserName

(R)OOHNH2NOO

CAS RN 56613-61-7

IUPAC Name: 2-amino-3-(4-nitrophenyl)propanoic acid
Synonyms: p-nitrophenylalanine P-nitrophenylalanine 4-nitrophenylalanine
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: GTVVZTAFGPQSPC-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)