missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-D-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog No. AAH2695903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2695903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2695903 Supplier Thermo Scientific Chemicals Supplier No. H2695903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56613-61-7
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.19
MDL Number MFCD00066016
InChI Key GTVVZTAFGPQSPC-UHFFFAOYNA-N
Synonym 4-nitro-d-phenylalanine, h-d-phe 4-no2-oh, p-nitro-d-phenylalanine, 2r-2-amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, 4-nitro, r-2-amino-3-4-nitrophenyl propanoic acid, d-4-nitrophe, ambotzhaa1044, pubchem13021, d-4-nitro phenylalanine
PubChem CID 679497
IUPAC Name 2-amino-3-(4-nitrophenyl)propanoic acid
SMILES NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Quantity 1 g
Formula Weight 210.19
Percent Purity 98%
Chemical Name or Material 4-Nitro-D-phenylalanine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-446-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only