missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)aniline, 98%

Catalog No. AAA1379803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1379803 1 g
AAA1379806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1379803 Supplier Thermo Scientific Chemicals Supplier No. A1379803

CAS: 393-11-3 | C7H5F3N2O2 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 393-11-3
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD00014717
InChI Key UTKUVRNVYFTEHF-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl aniline, 5-amino-2-nitrobenzotrifluoride, 3-amino-6-nitrobenzotrifluoride, benzenamine, 4-nitro-3-trifluoromethyl, 3-trifluoromethyl-4-nitroaniline, 2-nitro-5-amino benzotrifluoride, 3-trifluoromethyl-4-nitroanilide, flu-1, 3-amino-6-nitro benzotrifluoride
PubChem CID 94955
IUPAC Name 4-nitro-3-(trifluoromethyl)aniline
SMILES NC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
UN Number UN2811
Beilstein 2650702
Formula Weight 206.13
Percent Purity 98%
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)aniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 206-884-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only