missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2-(trifluoromethyl)benzoic acid, 97%

Catalog No. AAH6145203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6145203 1 g
AAH6145206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6145203 Supplier Thermo Scientific Chemicals Supplier No. H6145203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 320-37-6 | C8H4F3NO4 | 235.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 320-37-6
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.12
MDL Number MFCD04039246
InChI Key BPCKZQCTLCTDST-UHFFFAOYSA-N
Synonym 4-nitro-2-trifluoromethyl benzoic acid, 2-carboxy-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl-benzoic acid, 4-nitro-2-trifluoromethylbenzoic acid, benzoic acid, 4-nitro-2-trifluoromethyl, 4-nitro-2-trifluoromethyl benzoic acid, pubchem1342, ksc496i1p, rarechem al bo 1444, 2-trifluoromethyl-4-nitrobenzoic acid
PubChem CID 2782737
IUPAC Name 4-nitro-2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=C(C=C(C=C1)[N+]([O-])=O)C(F)(F)F

Specifications

Melting Point 128°C to 132°C
Color Yellow
Quantity 1 g
Formula Weight 235.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)benzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only