missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2,6-diphenylphenol 98.0+%, TCI America™

Catalog No. N06741G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06741G Supplier TCI America Supplier No. N06741G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2423-73-6
Molecular Formula C18H13NO3
Molecular Weight (g/mol) 291.306
MDL Number MFCD00075055
InChI Key YCXQKJXTGDYKIO-UHFFFAOYSA-N
Synonym 2′C-Hydroxy-5′C-nitro-m-terphenyl
PubChem CID 625985
IUPAC Name 4-nitro-2,6-diphenylphenol
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2O)C3=CC=CC=C3)[N+](=O)[O-]

Specifications

Melting Point 137°C
Color Yellow
Quantity 1 g
Formula Weight 291.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-2,6-diphenylphenol