missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-1-naphthylamine 98.0+%, TCI America™

$221.39 - $673.80

Chemical Identifiers

CAS 776-34-1
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00004026
InChI Key BVPJPRYNQHAOPQ-UHFFFAOYSA-N
Synonym 4-nitro-1-naphthylamine, 1-amino-4-nitronaphthalene, 1-naphthalenamine, 4-nitro, 1,4-nitronaphthylamine, 4-nitro-1-naphthamine, 1-naphthylamine, 4-nitro, 4-nitro-1-naphthalenamine, 4-nitro-naphthalen-1-ylamine, 4-nitro-alpha-naphthylamine, 4-nitronaphthylamine
PubChem CID 13057
IUPAC Name 4-nitronaphthalen-1-amine
SMILES C1=CC=C2C(=C1)C(=CC=C2[N+](=O)[O-])N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N02135G
View Documents
TCI America
N02135G
5 g
Each for $221.39
Only null left
 
N021325G
View Documents
TCI America
N021325G
25 g
Each for $673.80
Only null left
 
Description

Description

Specifications

Chemical Identifiers

776-34-1
188.186
BVPJPRYNQHAOPQ-UHFFFAOYSA-N
13057
C1=CC=C2C(=C1)C(=CC=C2[N+](=O)[O-])N
C10H8N2O2
MFCD00004026
4-nitro-1-naphthylamine, 1-amino-4-nitronaphthalene, 1-naphthalenamine, 4-nitro, 1,4-nitronaphthylamine, 4-nitro-1-naphthamine, 1-naphthylamine, 4-nitro, 4-nitro-1-naphthalenamine, 4-nitro-naphthalen-1-ylamine, 4-nitro-alpha-naphthylamine, 4-nitronaphthylamine
4-nitronaphthalen-1-amine

Specifications

776-34-1
C10H8N2O2
5 g
BVPJPRYNQHAOPQ-UHFFFAOYSA-N
4-nitronaphthalen-1-amine
13057
≥98.0% (GC)
4-Nitro-1-naphthylamine
195°C
MFCD00004026
4-nitro-1-naphthylamine, 1-amino-4-nitronaphthalene, 1-naphthalenamine, 4-nitro, 1,4-nitronaphthylamine, 4-nitro-1-naphthamine, 1-naphthylamine, 4-nitro, 4-nitro-1-naphthalenamine, 4-nitro-naphthalen-1-ylamine, 4-nitro-alpha-naphthylamine, 4-nitronaphthylamine
C1=CC=C2C(=C1)C(=CC=C2[N+](=O)[O-])N
188.186
188.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0327

RTECSNumber : QM4370000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'