missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-1-naphthylamine 98.0+%, TCI America™

Catalog No. N02135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02135G 5 g
N021325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02135G Supplier TCI America Supplier No. N02135G
Only null left

Chemical Identifiers

CAS 776-34-1
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00004026
InChI Key BVPJPRYNQHAOPQ-UHFFFAOYSA-N
Synonym 4-nitro-1-naphthylamine, 1-amino-4-nitronaphthalene, 1-naphthalenamine, 4-nitro, 1,4-nitronaphthylamine, 4-nitro-1-naphthamine, 1-naphthylamine, 4-nitro, 4-nitro-1-naphthalenamine, 4-nitro-naphthalen-1-ylamine, 4-nitro-alpha-naphthylamine, 4-nitronaphthylamine
PubChem CID 13057
IUPAC Name 4-nitronaphthalen-1-amine
SMILES C1=CC=C2C(=C1)C(=CC=C2[N+](=O)[O-])N

Specifications

Melting Point 195°C
Quantity 5 g
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-1-naphthylamine
EINECSNumber (4)-0327
RTECSNumber QM4370000
TSCA No