missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-1-naphthol 98.0+%, TCI America™
$346.71 - $1056.92
Chemical Identifiers
| CAS | 605-62-9 |
|---|---|
| Molecular Formula | C10H7NO3 |
| Molecular Weight (g/mol) | 189.17 |
| MDL Number | MFCD02179392 |
| InChI Key | AUIRNGLMBHIITH-UHFFFAOYSA-N |
| Synonym | 1-Hydroxy-4-nitronaphthalene |
| PubChem CID | 343829 |
| IUPAC Name | 4-nitronaphthalen-1-ol |
| SMILES | C1=CC=C2C(=C1)C(=CC=C2O)[N+](=O)[O-] |
Chemical Identifiers
| 605-62-9 | |
| 189.17 | |
| AUIRNGLMBHIITH-UHFFFAOYSA-N | |
| 343829 | |
| C1=CC=C2C(=C1)C(=CC=C2O)[N+](=O)[O-] |
| C10H7NO3 | |
| MFCD02179392 | |
| 1-Hydroxy-4-nitronaphthalene | |
| 4-nitronaphthalen-1-ol |
Specifications
| 605-62-9 | |
| Yellow | |
| MFCD02179392 | |
| 1-Hydroxy-4-nitronaphthalene | |
| C1=CC=C2C(=C1)C(=CC=C2O)[N+](=O)[O-] | |
| 189.17 | |
| 189.17 | |
| Crystalline Powder |
| 167°C | |
| C10H7NO3 | |
| 5 g | |
| AUIRNGLMBHIITH-UHFFFAOYSA-N | |
| 4-nitronaphthalen-1-ol | |
| 343829 | |
| ≥98.0% (T) | |
| 4-Nitro-1-naphthol |
Safety and Handling
TSCA : No