Learn More
BTX™ 4-needle Array Electrodes for BTX™ AgilePulse™ In Vivo
4-needle Array Electrodes are miniature needles designed to provide superior, highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse™ In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
Supplier: BTX™ 470043
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifications
| Gene and Vaccine Delivery | |
| Stainless Steel | |
| Electroporation Needle Array | |
| 0.3 mm | |
| 4-needle Array | |
| 3 mm | |
| 0.16 in. | |
| Fine Point | |
| Electrode Connector Cable |
| AgilePulse™ in Vivo Gene Delivery Electroporation System | |
| BTX Electroporation | |
| 0.01 in. | |
| Straight/Needle array | |
| 0.12 in. | |
| 1 Ea. | |
| 4 mm | |
| Sterile | |
| Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.