missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 98%

Catalog No. AAA1534206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1534206 5 g
AAA1534214 25 g
AAA1534222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1534206 Supplier Thermo Scientific Chemicals Supplier No. A1534206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 645-56-7 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 645-56-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002395
InChI Key KLSLBUSXWBJMEC-UHFFFAOYSA-N
Synonym 4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
PubChem CID 12580
ChEBI CHEBI:34434
IUPAC Name 4-propylphenol
SMILES CCCC1=CC=C(C=C1)O

Specifications

Melting Point 20°C to 22°C
Density 0.983
Boiling Point 232°C
Flash Point 106°C (222°F)
Refractive Index 1.523
Quantity 5 g
UN Number UN3145
Beilstein 1363429
Formula Weight 136.19
Percent Purity 98%
Chemical Name or Material 4-n-Propylphenol
Show More Show Less
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
EINECSNumber 211-446-2
RTECSNumber SM8610000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only