Learn More
4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
$238.32 - $238.32
Chemical Identifiers
| CAS | 134150-01-9 |
|---|---|
| Molecular Formula | C9H13BO2 |
| Molecular Weight (g/mol) | 164.01 |
| MDL Number | MFCD00859261 |
| InChI Key | WLCGYIWOKVWFLB-UHFFFAOYSA-N |
| Synonym | 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid |
| PubChem CID | 4100861 |
| IUPAC Name | (4-propylphenyl)boronic acid |
| SMILES | CCCC1=CC=C(C=C1)B(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5288406
|
Thermo Scientific Chemicals
H5288406 |
5 g |
Each for $238.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 134150-01-9 | |
| 164.01 | |
| WLCGYIWOKVWFLB-UHFFFAOYSA-N | |
| 4100861 | |
| CCCC1=CC=C(C=C1)B(O)O |
| C9H13BO2 | |
| MFCD00859261 | |
| 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid | |
| (4-propylphenyl)boronic acid |
Specifications
| 134150-01-9 | |
| C9H13BO2 | |
| 5 g | |
| WLCGYIWOKVWFLB-UHFFFAOYSA-N | |
| (4-propylphenyl)boronic acid | |
| 4100861 | |
| 98% |
| 106°C to 108°C | |
| MFCD00859261 | |
| 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid | |
| CCCC1=CC=C(C=C1)B(O)O | |
| 164.01 | |
| 164.01 | |
| 4-n-Propylbenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only