missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH5288406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5288406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5288406 Supplier Thermo Scientific Chemicals Supplier No. H5288406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134150-01-9 | Molecular Formula: C9H13BO2 | Molecular Weight: 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134150-01-9
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
MDL Number MFCD00859261
InChI Key WLCGYIWOKVWFLB-UHFFFAOYSA-N
Synonym 4-propylphenyl boronic acid, 4-propyl benzeneboronic acid, 4-propylphenylboronicacid, 4-n-propylphenylboronic acid, 4-n-propylbenzeneboronic acid, boronic acid, 4-propylphenyl, pubchem7892, acmc-209btz, 4-propylphenyboronic acid
PubChem CID 4100861
IUPAC Name (4-propylphenyl)boronic acid
SMILES CCCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 106°C to 108°C
Quantity 5 g
Formula Weight 164.01
Percent Purity 98%
Chemical Name or Material 4-n-Propylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only