missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octyloxyphenyl 4-Butylbenzoate 98.0+%, TCI America™
Supplier: TCI America B10921G
Specifications
| 4-n-Octyloxyphenyl 4-Butylbenzoate [Liquid Crystal] | |
| White | |
| MFCD00060084 | |
| 4-Butylbenzoic Acid 4-n-Octyloxyphenyl Ester | |
| CCCCCCCCOC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)CCCC | |
| 382.544 | |
| 382.54 | |
| Crystalline Powder |
| 42815-59-8 | |
| C25H34O3 | |
| 1 g | |
| DESAJRHSCPKXHK-UHFFFAOYSA-N | |
| (4-octoxyphenyl) 4-butylbenzoate | |
| 19013731 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 42815-59-8 | |
| 382.544 | |
| DESAJRHSCPKXHK-UHFFFAOYSA-N | |
| 19013731 | |
| CCCCCCCCOC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)CCCC |
| C25H34O3 | |
| MFCD00060084 | |
| 4-Butylbenzoic Acid 4-n-Octyloxyphenyl Ester | |
| (4-octoxyphenyl) 4-butylbenzoate |
Safety and Handling
TSCA : No