accessibility menu, dialog, popup

UserName

CAS RN 42815-59-8

H3COOOCH3

CAS RN 42815-59-8

IUPAC Name: 4-(octyloxy)phenyl 4-butylbenzoate
Synonyms: 4-(octyloxy)phenyl 4-butylbenzoate
Molecular Weight (g/mol): 382.54
Molecular Formula: C25H34O3
InChi Key: DESAJRHSCPKXHK-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=CC=C(OC(=O)C2=CC=C(CCCC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results