missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octyloxyphenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 60°C |
| Boiling Point | 127°C |
| Quantity | 5 g |
| Formula Weight | 222.33 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-n-Octyloxyphenol |
Safety and Handling
| EINECSNumber | (3)-3539 |
| TSCA | Yes |