missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Octylbenzoic Acid 97.0+%, TCI America™
$132.35 - $331.81
Chemical Identifiers
| CAS | 3575-31-3 |
|---|---|
| Molecular Formula | C15H22O2 |
| Molecular Weight (g/mol) | 234.339 |
| MDL Number | MFCD00042649 |
| InChI Key | ZQLDNJKHLQOJGE-UHFFFAOYSA-N |
| Synonym | 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984 |
| PubChem CID | 19147 |
| IUPAC Name | 4-octylbenzoic acid |
| SMILES | CCCCCCCCC1=CC=C(C=C1)C(=O)O |
Chemical Identifiers
| 3575-31-3 | |
| 234.339 | |
| ZQLDNJKHLQOJGE-UHFFFAOYSA-N | |
| 19147 | |
| CCCCCCCCC1=CC=C(C=C1)C(=O)O |
| C15H22O2 | |
| MFCD00042649 | |
| 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984 | |
| 4-octylbenzoic acid |
Specifications
| 3575-31-3 | |
| White | |
| MFCD00042649 | |
| 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984 | |
| CCCCCCCCC1=CC=C(C=C1)C(=O)O | |
| 234.339 | |
| 234.34 | |
| Crystalline Powder |
| 113°C | |
| C15H22O2 | |
| 1 g | |
| ZQLDNJKHLQOJGE-UHFFFAOYSA-N | |
| 4-octylbenzoic acid | |
| 19147 | |
| ≥97.0% (GC,T) | |
| 4-n-Octylbenzoic Acid |
Safety and Handling
EINECSNumber : (3)-3544
RTECSNumber : DH5945000
TSCA : Yes