Learn More
4-n-Octylbenzoic acid, 99%
CAS: 3575-31-3 | C15H22O2 | 234.339 g/mol
$120.04 - $517.23
Chemical Identifiers
| CAS | 3575-31-3 |
|---|---|
| Molecular Formula | C15H22O2 |
| Molecular Weight (g/mol) | 234.339 |
| MDL Number | MFCD00042649 |
| InChI Key | ZQLDNJKHLQOJGE-UHFFFAOYSA-N |
| Synonym | 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984 |
| PubChem CID | 19147 |
| IUPAC Name | 4-octylbenzoic acid |
| SMILES | CCCCCCCCC1=CC=C(C=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1077703
|
Thermo Scientific Chemicals
A1077703 |
1 g |
Each for $120.04
|
|
|||||
|
AAA1077706
|
Thermo Scientific Chemicals
A1077706 |
5 g |
Each for $517.23
|
|
|||||
Description
4-n-Octylbenzoic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-n-Octylbenzoic acid is used as a pharmaceutical intermediate.
Solubility
Soluble in methanol, hot hexane, hot ethanol and 2-propanol and toluene. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 3575-31-3 | |
| 234.339 | |
| ZQLDNJKHLQOJGE-UHFFFAOYSA-N | |
| 19147 | |
| CCCCCCCCC1=CC=C(C=C1)C(=O)O |
| C15H22O2 | |
| MFCD00042649 | |
| 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984 | |
| 4-octylbenzoic acid |
Specifications
| 3575-31-3 | |
| C15H22O2 | |
| 1 g | |
| 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984 | |
| ZQLDNJKHLQOJGE-UHFFFAOYSA-N | |
| 4-octylbenzoic acid | |
| 19147 | |
| 99% |
| 97°C to 98°C | |
| MFCD00042649 | |
| 1954607 | |
| Soluble in methanol,hot hexane,hot ethanol and 2-propanol and toluene. Insoluble in water. | |
| CCCCCCCCC1=CC=C(C=C1)C(=O)O | |
| 234.339 | |
| 234.34 | |
| 4-n-Octylbenzoic acid |
Safety and Handling
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302-H319-H335
EINECSNumber : 222-692-5
RTECSNumber : DH5945000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only