missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylbenzoic acid, 99%

Catalog No. AAA1077703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1077703 1 g
AAA1077706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1077703 Supplier Thermo Scientific Chemicals Supplier No. A1077703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3575-31-3 | C15H22O2 | 234.339 g/mol

4-n-Octylbenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Octylbenzoic acid is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol, hot hexane, hot ethanol and 2-propanol and toluene. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3575-31-3
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00042649
InChI Key ZQLDNJKHLQOJGE-UHFFFAOYSA-N
Synonym 4-n-octylbenzoic acid, benzoic acid, 4-octyl, p-octylbenzoic acid, para-octylbenzoic acid, benzoic acid, p-octyl, p-n-octyl benzoic acid, p-octyl benzoic acid, 4-n-octylbenzoicacid, p-n-octylbenzoic acid, maybridge1_001984
PubChem CID 19147
IUPAC Name 4-octylbenzoic acid
SMILES CCCCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 97°C to 98°C
Quantity 1 g
Beilstein 1954607
Solubility Information Soluble in methanol,hot hexane,hot ethanol and 2-propanol and toluene. Insoluble in water.
Formula Weight 234.34
Percent Purity 99%
Chemical Name or Material 4-n-Octylbenzoic acid
Hazard Category H302-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 222-692-5
RTECSNumber DH5945000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only