missing translation for 'onlineSavingsMsg'
Learn More

4-n-Nonylbenzeneboronic acid, 98+%

Catalog No. AAL1775306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1775306 5 g
AAL1775303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1775306 Supplier Thermo Scientific Chemicals Supplier No. L1775306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 256383-45-6 | C15H25BO2 | 248.173 g/mol

4-n-Nonylbenzeneboronic acid is a potent inhibitor of fatty acid amide hydrolase. It is also able to inhibit monoacylglycerol lipase (MAGL), which hydrolyzes 2-arachidonoyl glycerol (2-AG).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Nonylbenzeneboronic acid is a potent inhibitor of fatty acid amide hydrolase. It is also able to inhibit monoacylglycerol lipase (MAGL), which hydrolyzes 2-arachidonoyl glycerol (2-AG).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 256383-45-6
Molecular Formula C15H25BO2
Molecular Weight (g/mol) 248.173
MDL Number MFCD02093070
InChI Key VONVJOGSLHAKOX-UHFFFAOYSA-N
Synonym 4-n-nonylphenylboronic acid, 4-n-nonylbenzeneboronic acid, 4-nonylphenyl boronic acid, 4-n-nonylphenylboronicacid, 4-nonylphenyl boranediol, d0f1ud, 4-nonylbenzene boronic acid, b-4-nonylphenyl boronic acid, 4-n-nonyl benzeneboronic acid, 1-borono-4-non-1-yl benzene
PubChem CID 4589192
IUPAC Name (4-nonylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCCCCCCC)(O)O

Specifications

Melting Point 82°C to 85°C
Quantity 5 g
Beilstein 8215118
Solubility Information Insoluble in water.
Formula Weight 248.18
Percent Purity ≥98%
Chemical Name or Material 4-n-Nonylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only