CAS RN 256383-45-6
IUPAC Name:
(4-nonylphenyl)boronic acid
Synonyms:
4-nonylphenylboronic acid
Molecular Weight (g/mol):
248.17
Molecular Formula:
C15H25BO2
InChi Key:
VONVJOGSLHAKOX-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
Promotions Available
4-n-Nonylbenzeneboronic acid, 98+%
4-n-Nonylbenzeneboronic acid, 98+%
CAS: 256383-45-6 Molecular Formula: C15H25BO2 Molecular Weight (g/mol): 248.173 MDL Number: MFCD02093070 InChI Key: VONVJOGSLHAKOX-UHFFFAOYSA-N Synonym: 4-n-nonylphenylboronic acid,4-n-nonylbenzeneboronic acid,4-nonylphenyl boronic acid,4-n-nonylphenylboronicacid,4-nonylphenyl boranediol,d0f1ud,4-nonylbenzene boronic acid,b-4-nonylphenyl boronic acid,4-n-nonyl benzeneboronic acid,1-borono-4-non-1-yl benzene PubChem CID: 4589192 IUPAC Name: (4-nonylphenyl)boronic acid SMILES: B(C1=CC=C(C=C1)CCCCCCCCC)(O)O