missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Nonylaniline, 98%
CAS: 37529-29-6 | C15H25N | 219.37 g/mol
$67.08 - $646.10
Chemical Identifiers
| CAS | 37529-29-6 |
|---|---|
| Molecular Formula | C15H25N |
| Molecular Weight (g/mol) | 219.37 |
| MDL Number | MFCD00038370 |
| InChI Key | FDECURPHVMNAKO-UHFFFAOYSA-N |
| Synonym | p-nonylaniline, p-n-nonylaniline, 4-n-nonylaniline, p-nonyl aniline, p-nonylamiline, benzenamine, 4-nonyl, 1-amino-4-n-nonylbenzene |
| PubChem CID | 142170 |
| IUPAC Name | 4-nonylaniline |
| SMILES | CCCCCCCCCC1=CC=C(N)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3341403
|
Thermo Scientific Chemicals
H3341403 |
1 g |
Each for $67.08
|
|
|||||
|
AAH3341406
|
Thermo Scientific Chemicals
H3341406 |
5 g |
Each for $195.95
|
|
|||||
|
AAH3341414
|
Thermo Scientific Chemicals
H3341414 |
25 g |
Each for $646.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 37529-29-6 | |
| 219.37 | |
| FDECURPHVMNAKO-UHFFFAOYSA-N | |
| 142170 | |
| CCCCCCCCCC1=CC=C(N)C=C1 |
| C15H25N | |
| MFCD00038370 | |
| p-nonylaniline, p-n-nonylaniline, 4-n-nonylaniline, p-nonyl aniline, p-nonylamiline, benzenamine, 4-nonyl, 1-amino-4-n-nonylbenzene | |
| 4-nonylaniline |
Specifications
| 37529-29-6 | |
| >110°C (230°F) | |
| MFCD00038370 | |
| p-nonylaniline, p-n-nonylaniline, 4-n-nonylaniline, p-nonyl aniline, p-nonylamiline, benzenamine, 4-nonyl, 1-amino-4-n-nonylbenzene | |
| CCCCCCCCCC1=CC=C(N)C=C1 | |
| 219.37 | |
| 219.37 | |
| 4-n-Nonylaniline |
| 144°C (2 mmHg) | |
| C15H25N | |
| 1 g | |
| FDECURPHVMNAKO-UHFFFAOYSA-N | |
| 4-nonylaniline | |
| 142170 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only