Learn More
4-n-Nonylaniline, 98%
CAS: 37529-29-6 | C15H25N | 219.37 g/mol
Supplier: Thermo Scientific Chemicals H3341406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-n-Nonylaniline | |
| 144°C (2 mmHg) | |
| C15H25N | |
| 5 g | |
| FDECURPHVMNAKO-UHFFFAOYSA-N | |
| 4-nonylaniline | |
| 142170 | |
| 98% |
| 37529-29-6 | |
| >110°C (230°F) | |
| MFCD00038370 | |
| p-nonylaniline, p-n-nonylaniline, 4-n-nonylaniline, p-nonyl aniline, p-nonylamiline, benzenamine, 4-nonyl, 1-amino-4-n-nonylbenzene | |
| CCCCCCCCCC1=CC=C(N)C=C1 | |
| 219.37 | |
| 219.37 |
Chemical Identifiers
| 37529-29-6 | |
| 219.37 | |
| FDECURPHVMNAKO-UHFFFAOYSA-N | |
| 142170 | |
| CCCCCCCCCC1=CC=C(N)C=C1 |
| C15H25N | |
| MFCD00038370 | |
| p-nonylaniline, p-n-nonylaniline, 4-n-nonylaniline, p-nonyl aniline, p-nonylamiline, benzenamine, 4-nonyl, 1-amino-4-n-nonylbenzene | |
| 4-nonylaniline |
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only