missing translation for 'onlineSavingsMsg'
Learn More

4-(N,N-Dimethylamino)phenylboronic acid, 96%

Catalog No. p-115703
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC359235000 500 mg Glass bottle
AC359230025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC359235000 Supplier Thermo Scientific Chemicals Supplier No. 359235000

CAS: 28611-39-4 | C8H12BNO2 | 165.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28611-39-4
Molecular Formula C8H12BNO2
Molecular Weight (g/mol) 165.00
MDL Number MFCD01074642
InChI Key RIIPFHVHLXPMHQ-UHFFFAOYSA-N
Synonym 4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid, 4-dimethylaminophenyl boronic acid, 4-dimethylamino phenyl boranediol, 4-n,n-dimethylphenylboronic acid
PubChem CID 2734344
IUPAC Name [4-(dimethylamino)phenyl]boronic acid
SMILES CN(C)C1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 227°C
Color Blue-Gray or Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 165
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 4-(N, N-Dimethylamino)phenylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only