missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxybenzaldehyde, 98%

Catalog No. AAA1328722
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1328722 100 g
AAA1328706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1328722 Supplier Thermo Scientific Chemicals Supplier No. A1328722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5736-94-7 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5736-94-7
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00016615
InChI Key GWXUVWKBVROFDM-UHFFFAOYSA-N
Synonym 4-hexyloxy benzaldehyde, 4-hexyloxybenzaldehyde, 4-n-hexyloxybenzaldehyde, p-hexyloxybenzaldehyde, benzaldehyde, 4-hexyloxy, benzaldehyde, p-hexyloxy, p-hexyloxy benzaldehyde, p-hexoxybenzaldehyde, 4-hexyloxbenzaldehyde, p-n-hexyloxybenzaldehyde
PubChem CID 79816
IUPAC Name 4-hexoxybenzaldehyde
SMILES CCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.992
Boiling Point 149°C to 151°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 100 g
Beilstein 1912341
Sensitivity Air Sensitive
Formula Weight 206.29
Percent Purity 98%
Chemical Name or Material 4-n-Hexyloxybenzaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-251-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only