accessibility menu, dialog, popup

UserName

CAS RN 5736-94-7

IUPAC Name: 4-(hexyloxy)benzaldehyde
Synonyms: 4-(hexyloxy)benzaldehyde
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: GWXUVWKBVROFDM-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)