missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexyloxyaniline, 99%

Catalog No. p-7052800
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5586806 5 g
AAH5586814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5586806 Supplier Thermo Scientific Chemicals Supplier No. H5586806

CAS: 39905-57-2 | C12H19NO | 193.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39905-57-2
Molecular Formula C12H19NO
Molecular Weight (g/mol) 193.29
MDL Number MFCD00007868
InChI Key DJRKHTCUXRGYEU-UHFFFAOYSA-N
Synonym 4-hexyloxyaniline, 4-hexyloxy aniline, p-hexyloxyaniline, benzenamine, 4-hexyloxy, 4-n-hexyloxyaniline, p-hexyloxy aniline, 4-hexyloxy benzenamine, unii-vmx449yp3l, aniline, p-hexyloxy, vmx449yp3l
PubChem CID 38360
IUPAC Name 4-hexoxyaniline
SMILES CCCCCCOC1=CC=C(N)C=C1

Specifications

Melting Point 43°C to 45°C
Color White to Brown
Boiling Point 155°C to 158°C (5 mmHg)
Flash Point 113°C (235°F)
Quantity 5 g
Formula Weight 193.29
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-n-Hexyloxyaniline
Hazard Category H302+H312+H332-H315-H319-H335-H373
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 254-696-8
RTECSNumber BY2308000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only