missing translation for 'onlineSavingsMsg'
Learn More

4'-n-Hexylbiphenyl-4-carboxylic acid, 99%

Catalog No. AAB2190003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2190003 1 g
AAB2190006 5 g
AAB2190014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2190003 Supplier Thermo Scientific Chemicals Supplier No. B2190003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 59662-48-5 | C19H33O2 | 293.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59662-48-5
Molecular Formula C19H22O2
Molecular Weight (g/mol) 282.38
MDL Number MFCD00143232
InChI Key ROJCBWVSXWIGAL-UHFFFAOYSA-N
Synonym 4-4-hexylphenyl benzoic acid, 4-hexyl-4'-biphenylcarboxylic acid, 4-hexyl-4'-carboxybiphenyl, 4-n-hexylbiphenyl-4'-carboxylic acid, 4'-hexyl-1,1'-biphenyl-4-carboxylic acid, pubchem9069, acmc-209mez, 4'-hexylbiphenyl-4-carboxylic acid, 4-hexylbiphenyl-4'-carboxylic acid, 4'-hexyl-biphenyl-4-carboxylic acid
PubChem CID 13554588
IUPAC Name 4'-hexyl-[1,1'-biphenyl]-4-carboxylic acid
SMILES CCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 222°C to 224°C
Quantity 1 g
Formula Weight 282.38
Percent Purity 99%
Chemical Name or Material 4'-n-Hexylbiphenyl-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only