missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylbenzoic acid, 99%

Catalog No. AAA1416803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1416803 1 g
AAA1416806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1416803 Supplier Thermo Scientific Chemicals Supplier No. A1416803

CAS: 21643-38-9 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21643-38-9
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00013997
InChI Key CPEPWESLFZVUEP-UHFFFAOYSA-N
Synonym benzoic acid, 4-hexyl, 4-n-hexylbenzoic acid, p-hexylbenzoic acid, benzoic acid, p-hexyl, p-n-hexylbenzoic acid, 4n-hexylbenzoic acid, maybridge1_002028, para-n-hexyl benzoic acid, acmc-1chu5, ksc201s3p
PubChem CID 30725
IUPAC Name 4-hexylbenzoic acid
SMILES CCCCCCC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 97°C to 99°C (C-N)
Quantity 1 g
Beilstein 2046454
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 4-n-Hexylbenzoic acid
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 244-490-6
RTECSNumber DH1485000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only