missing translation for 'onlineSavingsMsg'
Learn More

4-n-Hexylaniline, 98%

Catalog No. AAH2629706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2629706 5 g
AAH2629714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2629706 Supplier Thermo Scientific Chemicals Supplier No. H2629706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 33228-45-4 | C12H19N | 177.291 g/mol

4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.

Chemical Identifiers

CAS 33228-45-4
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00007927
InChI Key OVEMTTZEBOCJDV-UHFFFAOYSA-N
Synonym p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh
PubChem CID 36382
IUPAC Name 4-hexylaniline
SMILES CCCCCCC1=CC=C(C=C1)N

Specifications

Density 0.919
Boiling Point 283°C to 285°C
Flash Point >110°C (230°F)
Refractive Index 1.525
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 177.29
Percent Purity 98%
Chemical Name or Material 4-n-Hexylaniline
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 251-409-8
RTECSNumber BY2305000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only