Learn More
4-n-Dodecylaniline, 95%
CAS: 104-42-7 | C18H31N | 261.453 g/mol
$104.86 - $315.99
Chemical Identifiers
| CAS | 104-42-7 |
|---|---|
| Molecular Formula | C18H31N |
| Molecular Weight (g/mol) | 261.453 |
| MDL Number | MFCD00007919 |
| InChI Key | KLPPPIIIEMUEGP-UHFFFAOYSA-N |
| Synonym | p-dodecylaniline, benzenamine, 4-dodecyl, p-dodecyl aniline, 4-dodecylphenylamine, 4-n-dodecylamine, p-n-dodecylaniline, paradodecyl aniline, acmc-20amjq, 4-dodecylaniline, ksc180i8p |
| PubChem CID | 7701 |
| IUPAC Name | 4-dodecylaniline |
| SMILES | CCCCCCCCCCCCC1=CC=C(C=C1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6552803
|
Thermo Scientific Chemicals
H6552803 |
1 g |
Each for $104.86
|
|
|||||
|
AAH6552806
|
Thermo Scientific Chemicals
H6552806 |
5 g |
Each for $315.99
|
|
|||||
Description
4-Dodecylaniline has been used: in the synthesis of sodium 4-dodecylphenylazosulfonate, to investigate the water solubilization of single-walled carbon nanotubes (SWNTs), in proton transfer step during molecular oxygen (O2) reduction by decamethylferrocene (DMFc).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Dodecylaniline has been used: in the synthesis of sodium 4-dodecylphenylazosulfonate, to investigate the water solubilization of single-walled carbon nanotubes (SWNTs), in proton transfer step during molecular oxygen (O2) reduction by decamethylferrocene (DMFc).
Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
| 104-42-7 | |
| 261.453 | |
| KLPPPIIIEMUEGP-UHFFFAOYSA-N | |
| 7701 | |
| CCCCCCCCCCCCC1=CC=C(C=C1)N |
| C18H31N | |
| MFCD00007919 | |
| p-dodecylaniline, benzenamine, 4-dodecyl, p-dodecyl aniline, 4-dodecylphenylamine, 4-n-dodecylamine, p-n-dodecylaniline, paradodecyl aniline, acmc-20amjq, 4-dodecylaniline, ksc180i8p | |
| 4-dodecylaniline |
Specifications
| 104-42-7 | |
| 220°C to 221°C (15 mmHg) | |
| C18H31N | |
| 1 g | |
| 2646305 | |
| KLPPPIIIEMUEGP-UHFFFAOYSA-N | |
| 4-dodecylaniline | |
| 7701 | |
| 95% | |
| 4-n-Dodecylaniline |
| 35°C to 39°C | |
| 150°C (302°F) | |
| MFCD00007919 | |
| UN3077 | |
| p-dodecylaniline, benzenamine, 4-dodecyl, p-dodecyl aniline, 4-dodecylphenylamine, 4-n-dodecylamine, p-n-dodecylaniline, paradodecyl aniline, acmc-20amjq, 4-dodecylaniline, ksc180i8p | |
| CCCCCCCCCCCCC1=CC=C(C=C1)N | |
| 261.453 | |
| 261.45 | |
| Solid |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 203-201-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only