accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 104-42-7

IUPAC Name: 4-dodecylaniline
Synonyms: P-dodecylaniline
Molecular Weight (g/mol): 261.45
Molecular Formula: C18H31N
InChi Key: KLPPPIIIEMUEGP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=C(N)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)